Please login to your online account to display your discounted pricing

2-Methyl-4-nitroaniline, 99%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  AC163955000

 View more versions of this product

Catalog No. AC163955000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Fine Needles
Assay ≥98.5%
Formula Weight 152.15
Color Yellow to khaki or brownish
Quantity 500g
Melting Point 129° to 133°C
Beilstein 12, 846
CAS Max % 100.0
MDL Number MFCD00007734
CAS 99-52-5
Synonym 2-Amino-5-nitrotoluene
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Infrared Spectrum Authentic
Assay Percent Range 99%
CAS Min % 98.5
Chemical Name or Material 2-Methyl-4-nitroaniline
Name Note 99%
Molecular Formula C7H8N2O2
Linear Formula CH3C6H3(NO2)NH2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Methyl-4-nitroaniline
Name Note 99%
CAS 99-52-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8N2O2
Linear Formula CH3C6H3(NO2)NH2
Formula Weight 152.15
MDL Number MFCD00007734
Synonym 2-Amino-5-nitrotoluene
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents