missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-4-nitroaniline, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 163951000

 View more versions of this product

Catalog No. AC163951000



Specifications

Specifications

2-Methyl-4-nitroaniline
Glass bottle
129°C to 133°C
99%
98.5
98.5% min. (GC)
CH3C6H3(NO2)NH2
12, 846
Solubility in water: insoluble. Other solubilities: soluble in hot diluted hydrochloric acid,soluble in ethanol
CC1=C(C=CC(=C1)[N+](=O)[O-])N
152.15
152.15
99%
Authentic
Brown or Khaki to Yellow
100g
99-52-5
100.0
C7H8N2O2
MFCD00007734
1-amino-2-methyl-4-nitrobenzene
XTTIQGSLJBWVIV-UHFFFAOYSA-N
2-methyl-4-nitroaniline
7441
Crystalline Powder or Fine Needles

Chemical Identifiers

99-52-5
152.15
XTTIQGSLJBWVIV-UHFFFAOYSA-N
7441
CC1=C(C=CC(=C1)[N+](=O)[O-])N
C7H8N2O2
MFCD00007734
1-amino-2-methyl-4-nitrobenzene
2-methyl-4-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-762-1

SDS Documents