missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Methyl-4'-nitroacetanilide, 97%

Manufacturer:  Alfa Aesar™ B2205406

 View more versions of this product

Catalog No. AAB2205406



Description

Description

Applications
2′-Methyl-4′-nitroacetanilide acts as the reactant in the synthesis of TNT isomers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, bases.
Specifications

Specifications

2'-Methyl-4'-nitroacetanilide
5g
C9H10N2O3
2'-methyl-4'-nitroacetanilide, 2-methyl-4-nitroacetanilide, 4'-nitro-o-acetotoluidide, acetamide, n-2-methyl-4-nitrophenyl, acetamide,n-2-methyl-4-nitrophenyl, acetyl-p-nitro-o-toluidine, maybridge1_001434, n-2-methyl-4-nitrophenyl acetamide, n1-2-methyl-4-nitrophenyl acetamide, o-acetotoluidide, 4'-nitro
CC(=O)NC1=CC=C(C=C1C)[N+]([O-])=O
194.19
194.19
198°C to 200°C
2719-15-5
MFCD00033896
JZEOVPGWIWSSAK-UHFFFAOYSA-N
N-(2-methyl-4-nitrophenyl)acetamide
75936
97%

Chemical Identifiers

2719-15-5
194.19
JZEOVPGWIWSSAK-UHFFFAOYSA-N
75936
CC(=O)NC1=CC=C(C=C1C)[N+]([O-])=O
C9H10N2O3
MFCD00033896
2'-methyl-4'-nitroacetanilide, 2-methyl-4-nitroacetanilide, 4'-nitro-o-acetotoluidide, acetamide, n-2-methyl-4-nitrophenyl, acetamide,n-2-methyl-4-nitrophenyl, acetyl-p-nitro-o-toluidine, maybridge1_001434, n-2-methyl-4-nitrophenyl acetamide, n1-2-methyl-4-nitrophenyl acetamide, o-acetotoluidide, 4'-nitro
N-(2-methyl-4-nitrophenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications