Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-4-n-propyl-1,3-oxathiane, cis + trans, 99%

Chemical Identifiers

Chemical Name or Material 2-Methyl-4-n-propyl-1,3-oxathiane
CAS 67715-80-4
Name Note cis + trans
Molecular Formula C8H16OS
Formula Weight 160.27
MDL Number MFCD00673203
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2263006

  SDS
alfa aesar™
B2263006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2263014

  SDS
alfa aesar™
B2263014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.27
Chemical Name or Material 2-Methyl-4-n-propyl-1,3-oxathiane
Name Note cis + trans
Assay 99%
Molecular Formula C8H16OS
MDL Number MFCD00673203
CAS 67715-80-4
UN Number UN1993
Health Hazard 1 Warning
Boiling Point 218° to 220°C
Quantity 5g
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.