missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-4(5)-nitroimidazole, TCI America™

$24.44 - $190.97

Chemical Identifiers

CAS 696-23-1
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.103
MDL Number MFCD00005191
InChI Key FFYTTYVSDVWNMY-UHFFFAOYSA-N
Synonym 2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
PubChem CID 12760
IUPAC Name 2-methyl-5-nitro-1H-imidazole
SMILES CC1=NC=C(N1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M092225G View Documents TCI America
M092225G
25g Each for $24.44
 
M0922500G View Documents TCI America
M0922500G
500g Each for $190.97
 
Specifications

Chemical Identifiers

696-23-1
127.103
FFYTTYVSDVWNMY-UHFFFAOYSA-N
12760
CC1=NC=C(N1)[N+](=O)[O-]
C4H5N3O2
MFCD00005191
2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
2-methyl-5-nitro-1H-imidazole

Specifications

Yellow
25g
C4H5N3O2
2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
CC1=NC=C(N1)[N+](=O)[O-]
127.103
127.10
≥99.0% (T)
253°C
696-23-1
MFCD00005191
FFYTTYVSDVWNMY-UHFFFAOYSA-N
2-methyl-5-nitro-1H-imidazole
12760
Crystal-Powder at 20°C
2-Methyl-4(5)-nitroimidazole
Safety and Handling

Safety and Handling

RTECSNumber : NI7550000

TSCA : Yes