Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-4,5-diphenylthiazole, 97%

$226.11 - $226.11

Chemical Identifiers

Chemical Name or Material 2-Methyl-4,5-diphenylthiazole
CAS 3755-83-7
Molecular Formula C16H13NS
Formula Weight 251.35g/mol
MDL Number MFCD00186396
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5178106

 
alfa aesar™
H51781-06
5g Each for $226.11
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material 2-Methyl-4,5-diphenylthiazole
CAS 3755-83-7
Assay 97%
Molecular Formula C16H13NS
Formula Weight 251.35g/mol
MDL Number MFCD00186396