missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole, 97%, Thermo Scientific™
Chemical Identifiers
CAS
857283-68-2
Molecular Formula
C16H20BNO2S
Molecular Weight (g/mol)
301.211
MDL Number
MFCD08060513
InChI Key
XCAUOIGPROQXRG-UHFFFAOYSA-N
Synonym
2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl thiazole, 2-methyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-1,3-thiazole, thiazole,2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
PubChem CID
18525749
IUPAC Name
2-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole
SMILES
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CSC(=N3)C
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
CAS
857283-68-2
Molecular Weight (g/mol)
301.211
InChI Key
XCAUOIGPROQXRG-UHFFFAOYSA-N
PubChem CID
18525749
SMILES
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CSC(=N3)C
Molecular Formula
C16H20BNO2S
MDL Number
MFCD08060513
Synonym
2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl thiazole, 2-methyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-1,3-thiazole, thiazole,2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
IUPAC Name
2-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole
Specifications
CAS
857283-68-2
CAS Max %
100.0
Packaging
Amber Glass Bottle
MDL Number
MFCD08060513
Synonym
2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl thiazole, 2-methyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-thiazole, 2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-1,3-thiazole, thiazole,2-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
SMILES
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CSC(=N3)C
Molecular Weight (g/mol)
301.211
Formula Weight
301.22
Chemical Name or Material
2-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole
CAS Min %
96.5
Assay Percent Range
97%
Molecular Formula
C16H20BNO2S
Quantity
250mg
InChI Key
XCAUOIGPROQXRG-UHFFFAOYSA-N
IUPAC Name
2-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole
PubChem CID
18525749
Percent Purity
97%
Safety and Handling
Safety and Handling
GHS H Statement: May cause respiratory irritation. Causes skin irritation. Harmful if swallowed. Causes serious eye irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'