Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-4-(2-pyridyl)thiazole, 97%

Chemical Identifiers

Chemical Name or Material 2-Methyl-4-(2-pyridyl)thiazole
CAS 14384-71-5
Formula Weight 176.24
MDL Number MFCD22666465
Beilstein 1565687
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5887804

  SDS
alfa aesar™
H5887804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5887809

  SDS
alfa aesar™
H5887809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.24
Melting Point 180° to 185°C
Chemical Name or Material 2-Methyl-4-(2-pyridyl)thiazole
Physical Form Powder
Assay 97%
MDL Number MFCD22666465
CAS 14384-71-5
Beilstein 1565687
Color Pale brown
Quantity 2g