missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-trifluoromethylaniline 99.0+%, TCI America™

$95.50 - $330.47

Chemical Identifiers

CAS 54396-44-0
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD00153216
InChI Key TWLDBACVSHADLI-UHFFFAOYSA-N
Synonym 2-methyl-3 trifluoromethyl aniline
PubChem CID 2735932
IUPAC Name 2-methyl-3-(trifluoromethyl)aniline
SMILES CC1=C(C=CC=C1N)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13281G View Documents Tci America
M13281G
1g Each for $95.50
 
M13285G View Documents Tci America
M13285G
5g Each for $330.47
 
Specifications

Chemical Identifiers

54396-44-0
175.154
TWLDBACVSHADLI-UHFFFAOYSA-N
2735932
CC1=C(C=CC=C1N)C(F)(F)F
C8H8F3N
MFCD00153216
2-methyl-3 trifluoromethyl aniline
2-methyl-3-(trifluoromethyl)aniline

Specifications

Undesignated
1g
C8H8F3N
2-methyl-3 trifluoromethyl aniline
CC1=C(C=CC=C1N)C(F)(F)F
175.154
175.15
≥99.0% (GC)
40°C
54396-44-0
MFCD00153216
TWLDBACVSHADLI-UHFFFAOYSA-N
2-methyl-3-(trifluoromethyl)aniline
2735932
Crystal-Powder at 20°C
2-Methyl-3-trifluoromethylaniline
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4077

TSCA : No