Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-(trifluoromethyl)phenol, 97%

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-(trifluoromethyl)phenol
Molecular Formula C8H7F3O
Formula Weight 176.14
MDL Number MFCD09832280
UN Number UN3261
Synonym 3-Hydroxy-2-methylbenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3158603

  SDS
alfa aesar™
H3158603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.14
UN Number UN3261
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)phenol
Melting Point 45° to 48°C
Assay 97%
Health Hazard 1 Danger
MDL Number MFCD09832280
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Synonym 3-Hydroxy-2-methylbenzotrifluoride
Molecular Formula C8H7F3O
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.