Learn More
2-Methyl-3-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H3155203
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Methyl-3-(trifluoromethyl)benzylamine | |
C9H10F3N | |
MFCD06213551 | |
UN2735 | |
2-methyl-3-trifluoromethyl phenyl methanamine, 2-methyl-3-trifluoromethyl benzylamine, 1-2-methyl-3-trifluoromethyl phenyl methanamine, 2-methyl-3-trifluoromethylbenzylamine, 2-methyl-3-trifluoromethyl phenyl methylamine | |
CC1=C(C=CC=C1C(F)(F)F)CN | |
189.181 | |
189.18 |
771572-43-1 | |
1.4812 | |
1g | |
Air Sensitive | |
WXYVXJSOJCMWGN-UHFFFAOYSA-N | |
[2-methyl-3-(trifluoromethyl)phenyl]methanamine | |
46737516 | |
97% |
Chemical Identifiers
771572-43-1 | |
189.181 | |
WXYVXJSOJCMWGN-UHFFFAOYSA-N | |
46737516 | |
CC1=C(C=CC=C1C(F)(F)F)CN |
C9H10F3N | |
MFCD06213551 | |
2-methyl-3-trifluoromethyl phenyl methanamine, 2-methyl-3-trifluoromethyl benzylamine, 1-2-methyl-3-trifluoromethyl phenyl methanamine, 2-methyl-3-trifluoromethylbenzylamine, 2-methyl-3-trifluoromethyl phenyl methylamine | |
[2-methyl-3-(trifluoromethyl)phenyl]methanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen