Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-(trifluoromethyl)benzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-(trifluoromethyl)benzonitrile
CAS 261952-02-7
Molecular Formula C9H6F3N
Formula Weight 185.15
MDL Number MFCD01631593
UN Number UN3439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2653303

  SDS
alfa aesar™
H2653303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2653306

  SDS
alfa aesar™
H2653306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.15
Assay 97%
Melting Point 28° to 31°C
Health Hazard 1 Danger
Quantity 1g
CAS 261952-02-7
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
MDL Number MFCD01631593
UN Number UN3439
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)benzonitrile
Molecular Formula C9H6F3N
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.