Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-(trifluoromethyl)aniline, 98%

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-(trifluoromethyl)aniline
CAS 54396-44-0
Molecular Formula C8H8F3N
Formula Weight 175.16
MDL Number MFCD00153216
UN Number UN2811
Beilstein 2832865
Synonym 3-Amino-2-methylbenzotrifluoride; 3-Trifluoromethyl-o-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2000903

  SDS
alfa aesar™
B2000903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2000906

  SDS
alfa aesar™
B2000906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.16
Molecular Formula C8H8F3N
MDL Number MFCD00153216
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Boiling Point 62° to 64°C (4mmHg)
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)aniline
Beilstein 2832865
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Synonym 3-Amino-2-methylbenzotrifluoride; 3-Trifluoromethyl-o-toluidine
Assay 98%
Melting Point 37° to 40°C
Flash Point 98°C (208°F)
CAS 54396-44-0
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.