missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-(trifluoromethyl)aniline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC263800010

 View more versions of this product

Catalog No. AC263800010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Methyl-3-(trifluoromethyl)aniline
Authentic
1.4825
White
1g
96.0
96% min. (GC)
2-methyl-3 trifluoromethyl aniline, 2-methyl-3-trifluoromethyl aniline, 2-methyl-3-trifluoromethyl benzenamine, 2-methyl-3-trifluoromethyl-phenylamine, 3-amino-2-methyl benzotrifluoride, 3-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-o-toluidine, benzenamine, 2-methyl-3-trifluoromethyl
CC1=C(C=CC=C1N)C(F)(F)F
175.16
175.16
97%
96°C
Glass Bottle
108.0°C to 110.0°C
39.0°C to 41.0°C
54396-44-0
100.0
C8H8F3N
TWLDBACVSHADLI-UHFFFAOYSA-N
2-methyl-3-(trifluoromethyl)aniline
2735932
Crystalline Powder

Chemical Identifiers

54396-44-0
175.16
2-methyl-3 trifluoromethyl aniline, 2-methyl-3-trifluoromethyl aniline, 2-methyl-3-trifluoromethyl benzenamine, 2-methyl-3-trifluoromethyl-phenylamine, 3-amino-2-methyl benzotrifluoride, 3-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-o-toluidine, benzenamine, 2-methyl-3-trifluoromethyl
2-methyl-3-(trifluoromethyl)aniline
C8H8F3N
TWLDBACVSHADLI-UHFFFAOYSA-N
2735932
CC1=C(C=CC=C1N)C(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 259-145-5

SDS Documents