Please login to your online account to display your discounted pricing

2-Methyl-3-(trifluoromethyl)aniline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-(trifluoromethyl)aniline
CAS 54396-44-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8F3N
Formula Weight 175.16
Synonym 3-(Trifluoromethyl)-o-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC263800050

 
thermo fisher scientific
AC263800050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC263800010

 
thermo fisher scientific
AC263800010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 175.16
Quantity 5g
Color White
Melting Point 37° to 42°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym 3-(Trifluoromethyl)-o-toluidine
Molecular Formula C8H8F3N
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)aniline
Assay Percent Range 97%
CAS 54396-44-0
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.