Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Methyl-3'-(trifluoromethyl)acetophenone, 97%

Chemical Identifiers

Chemical Name or Material 2'-Methyl-3'-(trifluoromethyl)acetophenone
Molecular Formula C10H9F3O
Formula Weight 202.17
MDL Number MFCD09832283
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3288803

  SDS
alfa aesar™
H3288803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3288806

  SDS
alfa aesar™
H3288806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.17
Molecular Formula C10H9F3O
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD09832283
Assay 97%
Quantity 1g
Chemical Name or Material 2'-Methyl-3'-(trifluoromethyl)acetophenone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.