missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-tetrahydrothiophenone, TCI America™

$175.70 - $534.73

Chemical Identifiers

CAS 13679-85-1
Molecular Formula C5H8OS
Molecular Weight (g/mol) 116.178
MDL Number MFCD00078280
InChI Key YMZZPMVKABUEBL-UHFFFAOYSA-N
Synonym 2-methyltetrahydrothiophen-3-one, dihydro-2-methyl-3 2h-thiophenone, 2-methyldihydrothiophen-3 2h-one, 3 2h-thiophenone, dihydro-2-methyl, 2-methyltetrahydrothiophene-3-one, 2-methyl-3-oxotetrahydrothiophene, 2-methyl-4,5-dihydro-3 2h-thiophenone, 2-methyl-3-tetrahydrothiophenone, 2-methyl tetrahydrothiophen-3-one, xi-dihydro-2-methyl-3 2h-thiophenone
PubChem CID 61664
ChEBI CHEBI:87506
IUPAC Name 2-methylthiolan-3-one
SMILES CC1C(=O)CCS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M18395G View Documents TCI America
M18395G
5g Each for $175.70
 
M183925G View Documents TCI America
M183925G
25g Each for $534.73
 
Specifications

Chemical Identifiers

13679-85-1
116.178
YMZZPMVKABUEBL-UHFFFAOYSA-N
61664
2-methylthiolan-3-one
C5H8OS
MFCD00078280
2-methyltetrahydrothiophen-3-one, dihydro-2-methyl-3 2h-thiophenone, 2-methyldihydrothiophen-3 2h-one, 3 2h-thiophenone, dihydro-2-methyl, 2-methyltetrahydrothiophene-3-one, 2-methyl-3-oxotetrahydrothiophene, 2-methyl-4,5-dihydro-3 2h-thiophenone, 2-methyl-3-tetrahydrothiophenone, 2-methyl tetrahydrothiophen-3-one, xi-dihydro-2-methyl-3 2h-thiophenone
CHEBI:87506
CC1C(=O)CCS1

Specifications

86°C
5g
C5H8OS
2-methyltetrahydrothiophen-3-one, dihydro-2-methyl-3 2h-thiophenone, 2-methyldihydrothiophen-3 2h-one, 3 2h-thiophenone, dihydro-2-methyl, 2-methyltetrahydrothiophene-3-one, 2-methyl-3-oxotetrahydrothiophene, 2-methyl-4,5-dihydro-3 2h-thiophenone, 2-methyl-3-tetrahydrothiophenone, 2-methyl tetrahydrothiophen-3-one, xi-dihydro-2-methyl-3 2h-thiophenone
CC1C(=O)CCS1
116.178
CHEBI:87506
Clear Liquid at 20°C
2-Methyl-3-tetrahydrothiophenone
Yellow
13679-85-1
MFCD00078280
YMZZPMVKABUEBL-UHFFFAOYSA-N
2-methylthiolan-3-one
61664
116.18
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes