missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-nitropyridine, TCI America™

$42.78 - $126.81

Chemical Identifiers

CAS 18699-87-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD04114137
InChI Key CCFGTKQIRWHYTB-UHFFFAOYSA-N
Synonym 2-methyl-3-nitro-pyridine, 3-nitro-2-picoline, 3-nitropicoline, 3-nitro-2-methylpyridine, pyridine, 2-methylnitro, pyridine, 2-methyl-3-nitro-9ci, nitropicoline, zlchem 137, acmc-209ep0, pyridine, 2-methyl-3-nitro
PubChem CID 329360
IUPAC Name 2-methyl-3-nitropyridine
SMILES CC1=C(C=CC=N1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25485G View Documents TCI America
M25485G
5g Each for $126.81
 
M25481G View Documents TCI America
M25481G
1g Each for $42.78
 
Specifications

Chemical Identifiers

18699-87-1
138.126
CCFGTKQIRWHYTB-UHFFFAOYSA-N
329360
CC1=C(C=CC=N1)[N+](=O)[O-]
C6H6N2O2
MFCD04114137
2-methyl-3-nitro-pyridine, 3-nitro-2-picoline, 3-nitropicoline, 3-nitro-2-methylpyridine, pyridine, 2-methylnitro, pyridine, 2-methyl-3-nitro-9ci, nitropicoline, zlchem 137, acmc-209ep0, pyridine, 2-methyl-3-nitro
2-methyl-3-nitropyridine

Specifications

86°C
5g
C6H6N2O2
2-methyl-3-nitro-pyridine, 3-nitro-2-picoline, 3-nitropicoline, 3-nitro-2-methylpyridine, pyridine, 2-methylnitro, pyridine, 2-methyl-3-nitro-9ci, nitropicoline, zlchem 137, acmc-209ep0, pyridine, 2-methyl-3-nitro
CC1=C(C=CC=N1)[N+](=O)[O-]
138.126
138.13
≥98.0% (GC)
Undesignated
18699-87-1
MFCD04114137
CCFGTKQIRWHYTB-UHFFFAOYSA-N
2-methyl-3-nitropyridine
329360
Crystal-Lump at 20°C
2-Methyl-3-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator