Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-nitrophenol, 96%

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-nitrophenol
CAS 5460-31-1
Molecular Formula C7H7NO3
Formula Weight 153.14
MDL Number MFCD00007241
UN Number UN2446
Beilstein 1946057
Synonym 2-Hydroxy-6-nitrotoluene; 3-Nitro-o-cresol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1300206

 
alfa aesar™
A1300206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1300203

 
alfa aesar™
A1300203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1300214

 
alfa aesar™
A1300214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.14
Chemical Name or Material 2-Methyl-3-nitrophenol
Assay 96%
Molecular Formula C7H7NO3
MDL Number MFCD00007241
CAS 5460-31-1
UN Number UN2446
Beilstein 1946057
Synonym 2-Hydroxy-6-nitrotoluene; 3-Nitro-o-cresol
Melting Point 148° to 152°C
Quantity 5g
Odor Phenol-like
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROCRESOLS, SOLID
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.