Please login to your online account to display your discounted pricing

2-Methyl-3-nitrobenzyl chloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-nitrobenzyl chloride
Name Note 97%
CAS 60468-54-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H8ClNO2
Linear Formula CH3C6H3(NO2)CH2Cl
Formula Weight 185.61
MDL Number MFCD00007165
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC187400010

  SDS
thermo fisher scientific
187400010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.61
Quantity 1g
TSCA TSCA
EINECS Number 262-249-3
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 1 GHS Signal Word: Danger
Name Note 97%
Linear Formula CH3C6H3(NO2)CH2Cl
Assay Percent Range 97%
Molecular Formula C8H8ClNO2
CAS 60468-54-4
Chemical Name or Material 2-Methyl-3-nitrobenzyl chloride
MDL Number MFCD00007165
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.