missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-nitrobenzotrifluoride, TCI America™

$166.53 - $501.13

Chemical Identifiers

CAS 6656-49-1
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.136
MDL Number MFCD00042322
InChI Key KQUQBPVYIURTNZ-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrobenzotrifluoride, 2-nitro-6-trifluoromethyltoluene, 2-methyl-1-nitro-3-trifluoromethyl benzene, 2-nitro-6-trifluoromethyl toluene, 2-methyl-3-trifluoromethyl nitrobenzene, benzene, 2-methyl-1-nitro-3-trifluoromethyl, alpha,alpha,alpha-trifluoro-3-nitro-o-xylene, 3-trifluoromethyl-2-methyl-1-nitrobenzene, pubchem4816, 2-methyl-3-nitro-benzotrifluoride
PubChem CID 81171
IUPAC Name 2-methyl-1-nitro-3-(trifluoromethyl)benzene
SMILES CC1=C(C=CC=C1[N+](=O)[O-])C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M12755G View Documents TCI America
M12755G
5g Each for $166.53
 
M127525G View Documents TCI America
M127525G
25g Each for $501.13
 
Specifications

Chemical Identifiers

6656-49-1
205.136
KQUQBPVYIURTNZ-UHFFFAOYSA-N
81171
CC1=C(C=CC=C1[N+](=O)[O-])C(F)(F)F
C8H6F3NO2
MFCD00042322
2-methyl-3-nitrobenzotrifluoride, 2-nitro-6-trifluoromethyltoluene, 2-methyl-1-nitro-3-trifluoromethyl benzene, 2-nitro-6-trifluoromethyl toluene, 2-methyl-3-trifluoromethyl nitrobenzene, benzene, 2-methyl-1-nitro-3-trifluoromethyl, alpha,alpha,alpha-trifluoro-3-nitro-o-xylene, 3-trifluoromethyl-2-methyl-1-nitrobenzene, pubchem4816, 2-methyl-3-nitro-benzotrifluoride
2-methyl-1-nitro-3-(trifluoromethyl)benzene

Specifications

Yellow
6656-49-1
MFCD00042322
KQUQBPVYIURTNZ-UHFFFAOYSA-N
2-methyl-1-nitro-3-(trifluoromethyl)benzene
81171
Clear Liquid at 20°C
2-Methyl-3-nitrobenzotrifluoride
5g
C8H6F3NO2
2-methyl-3-nitrobenzotrifluoride, 2-nitro-6-trifluoromethyltoluene, 2-methyl-1-nitro-3-trifluoromethyl benzene, 2-nitro-6-trifluoromethyl toluene, 2-methyl-3-trifluoromethyl nitrobenzene, benzene, 2-methyl-1-nitro-3-trifluoromethyl, alpha,alpha,alpha-trifluoro-3-nitro-o-xylene, 3-trifluoromethyl-2-methyl-1-nitrobenzene, pubchem4816, 2-methyl-3-nitro-benzotrifluoride
CC1=C(C=CC=C1[N+](=O)[O-])C(F)(F)F
205.136
205.14
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No