Please login to your online account to display your discounted pricing

2-Methyl-3-nitrobenzoic Acid 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-nitrobenzoic acid
Name Note 99%
CAS 1975-50-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7NO4
Linear Formula CH3C6H3(NO2)CO2H
Formula Weight 181.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160171000

  SDS
fisher scientific acros
160171000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 181.15
Quantity 100g
Color Light yellow
Beilstein 09, 471
EINECS Number 217-826-4
TSCA TSCA
Melting Point 182° to 186°C
CAS Max % 100.0
Synonym 3-Nitro-o-toluic acid
Infrared Spectrum Authentic
Packaging Plastic bottle
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methyl-3-nitrobenzoic acid
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H7NO4
Linear Formula CH3C6H3(NO2)CO2H
MDL Number MFCD00007160
CAS 1975-50-4