Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-nitrobenzoic acid, 99%

$81.58 - $278.61

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-nitrobenzoic acid
CAS 1975-50-4
Molecular Formula C8H7NO4
Formula Weight 181.15
MDL Number MFCD00007160
Beilstein 2050096
Synonym 3-Nitro-o-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1469422

  SDS
alfa aesar™
A1469422
100g Each for $81.58

AAA1469430

  SDS
alfa aesar™
A1469430
250g Each for $153.93

AAA1469436

  SDS
alfa aesar™
A1469436
500g Each for $278.61
Description & Specifications

Specifications

Formula Weight 181.15
Assay 99%
Molecular Formula C8H7NO4
MDL Number MFCD00007160
Beilstein 2050096
Synonym 3-Nitro-o-toluic acid
Health Hazard 1 Warning
Quantity 100g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Methyl-3-nitrobenzoic acid
TSCA TSCA
CAS 1975-50-4
Melting Point 182° to 186°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.