missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-nitrobenzoic acid, 99%

Manufacturer:  Alfa Aesar™ A1469436

 View more versions of this product

Catalog No. AAA1469436



Specifications

Specifications

2-Methyl-3-nitrobenzoic acid
500g
C8H7NO4
2050096
YPQAFWHSMWWPLX-UHFFFAOYSA-N
181.147
181.15
182°C to 186°C
1975-50-4
MFCD00007160
2-methyl-3-nitrobenzoic acid, 3-nitro-o-toluic acid, benzoic acid, 2-methyl-3-nitro, 2-methyl-3-nitrobenzoicacid, unii-0si17d349b, 3-nitro-2-methyl benzoic acid, 3-nitro-2-methylbenzoic acid, 2-methyl-3-nitro-benzoic acid, benzoic acid,2-methyl-3-nitro, pubchem4522
CC1=C(C=CC=C1[N+](=O)[O-])C(=O)O
16096
99%

Chemical Identifiers

1975-50-4
181.147
YPQAFWHSMWWPLX-UHFFFAOYSA-N
16096
C8H7NO4
MFCD00007160
2-methyl-3-nitrobenzoic acid, 3-nitro-o-toluic acid, benzoic acid, 2-methyl-3-nitro, 2-methyl-3-nitrobenzoicacid, unii-0si17d349b, 3-nitro-2-methyl benzoic acid, 3-nitro-2-methylbenzoic acid, 2-methyl-3-nitro-benzoic acid, benzoic acid,2-methyl-3-nitro, pubchem4522
CC1=C(C=CC=C1[N+](=O)[O-])C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents