Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-nitroanisole, 98+%

$69.27 - $229.35

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-nitroanisole
CAS 4837-88-1
Molecular Formula C8H9NO3
Formula Weight 167.16
MDL Number MFCD00007159
Beilstein 2328599
Synonym 3-Methoxy-2-methyl-1-nitrobenzene; 6-Methoxy-2-nitrotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1879203

 
alfa aesar™
L1879203
1g Each for $69.27

AAL1879206

 
alfa aesar™
L1879206
5g Each for $229.35
Description & Specifications

Specifications

Formula Weight 167.16
Chemical Name or Material 2-Methyl-3-nitroanisole
CAS 4837-88-1
Beilstein 2328599
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Molecular Formula C8H9NO3
MDL Number MFCD00007159
Synonym 3-Methoxy-2-methyl-1-nitrobenzene; 6-Methoxy-2-nitrotoluene
Health Hazard 1 Danger
Melting Point 52° to 54°C
Boiling Point 94° to 96°C (0.5mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.