Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-nitroaniline, 98%

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-nitroaniline
CAS 603-83-8
Molecular Formula C7H8N2O2
Formula Weight 152.15
MDL Number MFCD00007731
UN Number UN2660
Beilstein 388393
Synonym 2-Amino-6-nitrotoluene; 3-Nitro-o-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1034722

  SDS
alfa aesar™
A1034722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1034736

  SDS
alfa aesar™
A1034736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.15
Chemical Name or Material 2-Methyl-3-nitroaniline
Assay 98%
Molecular Formula C7H8N2O2
MDL Number MFCD00007731
CAS 603-83-8
UN Number UN2660
Beilstein 388393
Synonym 2-Amino-6-nitrotoluene; 3-Nitro-o-toluidine
Health Hazard 1 Danger
Melting Point 88° to 91°C
Boiling Point 305°C
Quantity 100g
Odor Characteristic
Health Hazard 2 GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P280h-P273-P309-P310-P302+P352
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)
RTECS Number XU8200000
Safety and Handling

Danger

GHS P Statement
P280h-P273-P309-P310-P302+P352
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.