Learn More
2-Methyl-3-nitroaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 152020250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methyl-3-nitroaniline | |
96.0 | |
87°C to 91°C | |
305°C | |
96% min. (HPLC) | |
C7H8N2O2 | |
MFCD00007731 | |
12, 848 | |
Solubility in water: insoluble. Other solubilities: freely soluble in ethanol and ether | |
CC1=C(N)C=CC=C1[N+]([O-])=O | |
152.15 | |
152.15 | |
Crystalline Powder |
603-83-8 | |
100.0 | |
Green-Yellow to Yellow | |
Authentic | |
Glass bottle | |
CH3C6H3(NO2)NH2 | |
25 g | |
2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine | |
HFCFJYRLBAANKN-UHFFFAOYSA-N | |
2-methyl-3-nitroaniline | |
11783 | |
97% |
Chemical Identifiers
603-83-8 | |
152.15 | |
HFCFJYRLBAANKN-UHFFFAOYSA-N | |
11783 | |
CC1=C(N)C=CC=C1[N+]([O-])=O |
C7H8N2O2 | |
MFCD00007731 | |
2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine | |
2-methyl-3-nitroaniline |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 210-059-6
RUO – Research Use Only