Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-n-propylpyrazine, 99%

$67.81 - $754.89

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-n-propylpyrazine
CAS 15986-80-8
Molecular Formula C8H12N2
Formula Weight 136.2
MDL Number MFCD00010096
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2201306

  SDS
alfa aesar™
B2201306
5g Each for $67.81

AAB2201314

  SDS
alfa aesar™
B2201314
25g Each for $244.09

AAB2201322

  SDS
alfa aesar™
B2201322
100g Each for $754.89
Description & Specifications

Specifications

Formula Weight 136.2
Assay 99%
MDL Number MFCD00010096
CAS 15986-80-8
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 189° to 190°C
Chemical Name or Material 2-Methyl-3-n-propylpyrazine
Molecular Formula C8H12N2
Health Hazard 1 Warning
Flash Point 68°C (156°F)
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
TSCA TSCA
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.