missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-(methylthio)pyrazine, 99%

$78.09 - $1,219.18

Chemical Identifiers

CAS 2882-20-4
Molecular Formula C6H8N2S
Formula Weight 140.21
MDL Number MFCD00014610
Synonym 2-Methylmercapto-3-methylpyrazine; 2-Methyl-3-(methylmercapto)pyrazine
Chemical Name or Material 2-Methyl-3- (methylthio)pyrazine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1195703 SDS Alfa Aesar™
A1195703
1g Each for $78.09
AAA1195706 SDS Alfa Aesar™
A1195706
5g Each for $308.16
AAA1195714 SDS Alfa Aesar™
A1195714
25g Each for $1,219.18
Specifications Description & Specifications

Chemical Identifiers

2882-20-4
140.21
2-Methylmercapto-3-methylpyrazine; 2-Methyl-3-(methylmercapto)pyrazine
C6H8N2S
MFCD00014610
2-Methyl-3- (methylthio)pyrazine

Specifications

2882-20-4
140.21
2-Methylmercapto-3-methylpyrazine; 2-Methyl-3-(methylmercapto)pyrazine
1g
2-Methyl-3- (methylthio)pyrazine
C6H8N2S
MFCD00014610
213° to 214°C
99°C (210°F)
99%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA