missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-(methylthio)furan, TCI America™

$114.58 - $346.81

Chemical Identifiers

CAS 63012-97-5
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.189
MDL Number MFCD03701635
InChI Key OQVAOEIMSKZGAL-UHFFFAOYSA-N
PubChem CID 526618
IUPAC Name 2-methyl-3-methylsulfanylfuran
SMILES CC1=C(C=CO1)SC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M183825G View Documents TCI America
M183825G
25g Each for $346.81
 
M18385G View Documents TCI America
M18385G
5g Each for $114.58
 
Specifications

Chemical Identifiers

63012-97-5
128.189
OQVAOEIMSKZGAL-UHFFFAOYSA-N
2-methyl-3-methylsulfanylfuran
C6H8OS
MFCD03701635
526618
CC1=C(C=CO1)SC

Specifications

132°C
25g
C6H8OS
1993
CC1=C(C=CO1)SC
128.189
128.19
≥98.0% (GC)
Yellow
63012-97-5
MFCD03701635
OQVAOEIMSKZGAL-UHFFFAOYSA-N
2-methyl-3-methylsulfanylfuran
526618
Clear Liquid at 20°C
2-Methyl-3-(methylthio)furan
Safety and Handling

Safety and Handling

TSCA : No