missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-(methyldithio)furan, TCI America™

$48.89 - $143.61

Chemical Identifiers

CAS 65505-17-1
Molecular Formula C6H8OS2
Molecular Weight (g/mol) 160.249
MDL Number MFCD00038643
InChI Key SRUTWBWLFKSTIS-UHFFFAOYSA-N
Synonym Methyl 2-Methyl-3-furyl Disulfide
PubChem CID 47649
IUPAC Name 2-methyl-3-(methyldisulfanyl)furan
SMILES CC1=C(C=CO1)SSC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M18371G View Documents TCI America
M18371G
1g Each for $48.89
 
M18375G View Documents TCI America
M18375G
5g Each for $143.61
 
Specifications

Chemical Identifiers

65505-17-1
160.249
SRUTWBWLFKSTIS-UHFFFAOYSA-N
47649
CC1=C(C=CO1)SSC
C6H8OS2
MFCD00038643
Methyl 2-Methyl-3-furyl Disulfide
2-methyl-3-(methyldisulfanyl)furan

Specifications

37°C
1g
C6H8OS2
2810
SRUTWBWLFKSTIS-UHFFFAOYSA-N
2-methyl-3-(methyldisulfanyl)furan
47649
Clear Liquid at 20°C
2-Methyl-3-(methyldithio)furan
Yellow
65505-17-1
MFCD00038643
Methyl 2-Methyl-3-furyl Disulfide
CC1=C(C=CO1)SSC
160.249
160.25
≥98.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : JO1975000

TSCA : No

Recommended Storage : Refrigerator