Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-furoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-furoyl chloride
CAS 5555-00-0
Molecular Formula C6H5ClO2
Formula Weight 144.56g/mol
MDL Number MFCD03421418
UN Number UN3265
Synonym 2-Methylfuran-3-carbonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2720603

 
alfa aesar™
H27206-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 59° to 61°C (12mmHg)
Quantity 1g
Flash Point 76°C (169°F)
Chemical Name or Material 2-Methyl-3-furoyl chloride
CAS 5555-00-0
Assay 97%
Molecular Formula C6H5ClO2
Formula Weight 144.56g/mol
MDL Number MFCD03421418
UN Number UN3265
Synonym 2-Methylfuran-3-carbonyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

2° to 8°C