Please login to your online account to display your discounted pricing

2-Methyl-3-furoic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  330150010

 View more versions of this product

Catalog No. AC330150010

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-furoic acid
Color Index No. 2-Methyl-3-furoic acid
Color Light brown
Melting Point 97° to 102°C
Quantity 1g
CAS 6947-94-0
CAS Min % 97.5
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Needles and Chunks
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6 H6 O3
Formula Weight 126.11
MDL Number MFCD00092314

Specifications

Chemical Name or Material 2-Methyl-3-furoic acid
CAS 6947-94-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6 H6 O3
Formula Weight 126.11
MDL Number MFCD00092314
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents