Please login to your online account to display your discounted pricing

2-Methyl-3-furoic acid, 98%, ACROS Organics™

$188.48 - $188.48

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-furoic acid
CAS 6947-94-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6 H6 O3
Formula Weight 126.11
MDL Number MFCD00092314
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC330150010

  SDS
thermo fisher scientific
330150010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC330150050

  SDS
thermo fisher scientific
330150050
Glass bottle 5g Each for $188.48
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Needles and Chunks
Formula Weight 126.11
Quantity 1g
Color Light brown
Melting Point 97° to 102°C
Color Index No. 2-Methyl-3-furoic acid
CAS Max % 100.0
Chemical Name or Material 2-Methyl-3-furoic acid
Molecular Formula C6 H6 O3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Packaging Glass bottle
MDL Number MFCD00092314
Infrared Spectrum Authentic
CAS 6947-94-0
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.