missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-furanthiol 95.0+%, TCI America™

$115.22 - $339.05

Chemical Identifiers

CAS 28588-74-1
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.162
MDL Number MFCD00010280
InChI Key RUYNUXHHUVUINQ-UHFFFAOYSA-N
Synonym 2-methyl-3-furanthiol, 3-furanthiol, 2-methyl, 2-methyl-3-mercaptofuran, 2-methyl-3-furanethiol, 2-methyl-3-furylthiol, 2-methyl-furan-3-thiol, 3-mercapto-2-methylfuran, 2-methyl-3-furylmercaptan, 2-methyl-3-sulfanylfuran, unii-n21rw1n179
PubChem CID 34286
IUPAC Name 2-methylfuran-3-thiol
SMILES CC1=C(C=CO1)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M18471G View Documents Tci America
M18471G
1g Each for $115.22
 
M18475G View Documents Tci America
M18475G
5g Each for $339.05
 
Specifications

Chemical Identifiers

28588-74-1
114.162
RUYNUXHHUVUINQ-UHFFFAOYSA-N
34286
CC1=C(C=CO1)S
C5H6OS
MFCD00010280
2-methyl-3-furanthiol, 3-furanthiol, 2-methyl, 2-methyl-3-mercaptofuran, 2-methyl-3-furanethiol, 2-methyl-3-furylthiol, 2-methyl-furan-3-thiol, 3-mercapto-2-methylfuran, 2-methyl-3-furylmercaptan, 2-methyl-3-sulfanylfuran, unii-n21rw1n179
2-methylfuran-3-thiol

Specifications

60°C
1g
C5H6OS
1228
RUYNUXHHUVUINQ-UHFFFAOYSA-N
2-methylfuran-3-thiol
34286
Clear Liquid at 20°C
2-Methyl-3-furanthiol
Undesignated
28588-74-1
MFCD00010280
2-methyl-3-furanthiol, 3-furanthiol, 2-methyl, 2-methyl-3-mercaptofuran, 2-methyl-3-furanethiol, 2-methyl-3-furylthiol, 2-methyl-furan-3-thiol, 3-mercapto-2-methylfuran, 2-methyl-3-furylmercaptan, 2-methyl-3-sulfanylfuran, unii-n21rw1n179
CC1=C(C=CO1)S
114.162
114.16
≥95.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : LU6235000

TSCA : Yes

Recommended Storage : Refrigerator