missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-furanethiol, ca.90%, balance oxidized compound, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 275390010

 View more versions of this product

Catalog No. AC275390010



Specifications

Specifications

2-Methyl-3-furanethiol
37°C
Glass bottle
1.145
Orange to Pink
28588-74-1
100.0
C5H6OS
2-methyl-3-furanethiol, 2-methyl-3-furanthiol, 2-methyl-3-furylmercaptan, 2-methyl-3-furylthiol, 2-methyl-3-mercaptofuran, 2-methyl-3-sulfanylfuran, 2-methyl-furan-3-thiol, 3-furanthiol, 2-methyl, 3-mercapto-2-methylfuran, unii-n21rw1n179
RUYNUXHHUVUINQ-UHFFFAOYSA-N
2-methylfuran-3-thiol
34286
Liquid
1.1450g/mL
Authentic
1.5100 to 1.55
57°C to 60°C (44.0mmHg)
1g
88.0
88% min. (GC)
MFCD00010280
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
CC1=C(C=CO1)S
114.17
114.17
ca. 9%

Chemical Identifiers

28588-74-1
114.17
RUYNUXHHUVUINQ-UHFFFAOYSA-N
34286
CC1=C(C=CO1)S
C5H6OS
MFCD00010280
2-methyl-3-furanethiol, 2-methyl-3-furanthiol, 2-methyl-3-furylmercaptan, 2-methyl-3-furylthiol, 2-methyl-3-mercaptofuran, 2-methyl-3-sulfanylfuran, 2-methyl-furan-3-thiol, 3-furanthiol, 2-methyl, 3-mercapto-2-methylfuran, unii-n21rw1n179
2-methylfuran-3-thiol
Safety and Handling

Safety and Handling

GHS H Statement:
Fatal if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 249-094-7

Recommended Storage : important note: this substance readily oxidizes in air to form bis(2-methyl-3-fyryl)disulphide

SDS Documents