Please login to your online account to display your discounted pricing

2-Methyl-3-furanethiol, ca.90%, balance oxidized compound, ACROS Organics™

$91.62 - $91.62

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-furanethiol
CAS 28588-74-1
CAS Min % 88.0
CAS Max % 100.0
Assay ≥88%
Assay Percent Range ca.90%
Molecular Formula C5H6OS
Formula Weight 114.17
MDL Number MFCD00010280
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC275390050

  SDS
thermo fisher scientific
AC275390050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC275390010

  SDS
thermo fisher scientific
275390010
Glass bottle 1g Each for $91.62
Description & Specifications

Specifications

Assay ≥88%
Physical Form Liquid
Formula Weight 114.17
Quantity 5g
Color Clear to hazy light pink to light orange
Density 1.1450g/mL
CAS Max % 100.0
CAS Min % 88.0
Storage Information important note: this substance readily oxidizes in air to form bis(2-methyl-3-fyryl)disulphide
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C5H6OS
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methyl-3-furanethiol
CAS 28588-74-1
Refractive Index 1.5100 to 1.5500 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range ca.90%
MDL Number MFCD00010280
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Flammable liquid and vapor.