missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-3-butenenitrile 75.0+%, TCI America™
Supplier: TCI America M084025ML
Specifications
2-Methyl-3-butenenitrile | |
Yellow | |
C5H7N | |
25 mL | |
3-Cyano-1-butene, 1-Methyl-2-propenyl Cyanide | |
CC(C=C)C#N | |
81.118 | |
81.12 | |
Liquid |
16529-56-9 | |
124°C | |
MFCD00042634 | |
3273 | |
WBAXCOMEMKANRN-UHFFFAOYSA-N | |
2-methylbut-3-enenitrile | |
27909 | |
≥75.0% (GC) |
Chemical Identifiers
16529-56-9 | |
81.118 | |
WBAXCOMEMKANRN-UHFFFAOYSA-N | |
27909 | |
CC(C=C)C#N |
C5H7N | |
MFCD00042634 | |
3-Cyano-1-butene, 1-Methyl-2-propenyl Cyanide | |
2-methylbut-3-enenitrile |
Safety and Handling
TSCA : Yes