Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-buten-2-ol, 98%

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-buten-2-ol
CAS 115-18-4
Molecular Formula C5H10O
Formula Weight 86.13
MDL Number MFCD00004470
UN Number UN1987
Beilstein 1698263
Synonym Dimethyl vinyl carbinol; 3-Hydroxy-3-methyl-1-butene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB20790AE

 
alfa aesar™
B20790AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB20790AP

 
alfa aesar™
B20790AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 86.13
Chemical Name or Material 2-Methyl-3-buten-2-ol
TSCA TSCA
Assay 98%
Molecular Formula C5H10O
MDL Number MFCD00004470
CAS 115-18-4
UN Number UN1987
Beilstein 1698263
Synonym Dimethyl vinyl carbinol; 3-Hydroxy-3-methyl-1-butene
Melting Point −28°C
Boiling Point 98° to 99°C
Flash Point 13°C (55°F)
Health Hazard 1 Danger
Odor Like Alcohol
Quantity 100mL
Health Hazard 2 GHS H Statement
H225-H301-H319-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
RTECS Number EM9472000
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H319-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.