Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-3-[(3,4-methylenedioxy)phenyl]propionic acid

$81.22 - $232.04

Chemical Identifiers

Chemical Name or Material 2-Methyl-3-[(3,4-methylenedioxy)phenyl]propionic acid
CAS 77269-66-0
Molecular Formula C11H12O4
Formula Weight 208.21
MDL Number MFCD06203229
Synonym alpha-Methyl-1,3-benzodioxole-5-propionic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAH2792403

  SDS
alfa aesar™
H2792403
Each for $81.22

AAH2792406

  SDS
alfa aesar™
H2792406
Each for $232.04
Description & Specifications

Specifications

Formula Weight 208.21
Chemical Name or Material 2-Methyl-3-[(3,4-methylenedioxy)phenyl]propionic acid
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD06203229
Melting Point 77° to 80°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym alpha-Methyl-1,3-benzodioxole-5-propionic acid
Health Hazard 1 Warning
CAS 77269-66-0
Molecular Formula C11H12O4

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.