missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-(2-furyl)propenal, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1012518

 View more versions of this product

Catalog No. AAA1012518



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-3- (2-furyl)propenal
1.097
>100°C (212°F)
1.614
50 g
2-methyl-3-2-furyl propenal, alpha-methylfurylacrolein, 2-methyl-3-2-furyl acrolein, furfurylidene-2-propanal, furfurylidine-2-propanal, 2-methyl-3-furylacrolein, alpha-methyl-2-furanacrolein, 2-furfurylidenepropionaldehyde, 3-2-furyl methacrylaldehyde, 2-methyl-3 2-furyl acrolein
CC(=CC1=CC=CO1)C=O
136.15
136.15
874-66-8
280°C to 281°C
C8H8O2
MFCD00063240
2438
ZNBXZUKDRRRQJK-FNORWQNLSA-N
(E)-3-(furan-2-yl)-2-methylprop-2-enal
6210220
97%

Chemical Identifiers

874-66-8
136.15
ZNBXZUKDRRRQJK-FNORWQNLSA-N
6210220
CC(=CC1=CC=CO1)C=O
C8H8O2
MFCD00063240
2-methyl-3-2-furyl propenal, alpha-methylfurylacrolein, 2-methyl-3-2-furyl acrolein, furfurylidene-2-propanal, furfurylidine-2-propanal, 2-methyl-3-furylacrolein, alpha-methyl-2-furanacrolein, 2-furfurylidenepropionaldehyde, 3-2-furyl methacrylaldehyde, 2-methyl-3 2-furyl acrolein
(E)-3-(furan-2-yl)-2-methylprop-2-enal
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 212-866-9

RTECSNumber : LT8530600

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only