missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2H-indazole-4-boronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445140010

 View more versions of this product

Catalog No. AC445140010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Methyl-2H-indazole-4-boronic acid pinacol ester
145°C to 148°C
885698-95-3
C14H19BN2O2
YQMBOMPXHDRRIM-UHFFFAOYSA-N
44118254
97%
Glass Bottle
1g
96% min. (GC)
2-methyl-1h-indazole-4-boronic acid pinacol ester, 2-methyl-2h-indazol-4-yl boronic acid pinacol ester, 2-methyl-2h-indazole-4-boronic acid pinacol ester, 2-methyl-2h-indazole-4-boronic acid, pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-indazole, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-indazole, 2-methylindazole-4-boronic acid pinacol ester, yqmbompxhdrrim-uhfffaoysa-n
258.13
258.13

Chemical Identifiers

885698-95-3
258.13
2-methyl-1h-indazole-4-boronic acid pinacol ester, 2-methyl-2h-indazol-4-yl boronic acid pinacol ester, 2-methyl-2h-indazole-4-boronic acid pinacol ester, 2-methyl-2h-indazole-4-boronic acid, pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-indazole, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-indazole, 2-methylindazole-4-boronic acid pinacol ester, yqmbompxhdrrim-uhfffaoysa-n
C14H19BN2O2
YQMBOMPXHDRRIM-UHFFFAOYSA-N
44118254
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents