missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2H-indazole-4-boronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 885698-95-3
Molecular Formula C14H19BN2O2
Molecular Weight (g/mol) 258.13
InChI Key YQMBOMPXHDRRIM-UHFFFAOYSA-N
Synonym 2-methyl-1h-indazole-4-boronic acid pinacol ester
PubChem CID 44118254
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445140010 SDS View Documents Thermo Fisher Scientific
AC445140010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445140050 SDS View Documents Thermo Fisher Scientific
AC445140050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

885698-95-3
258.13
2-methyl-1h-indazole-4-boronic acid pinacol ester
C14H19BN2O2
YQMBOMPXHDRRIM-UHFFFAOYSA-N
44118254

Specifications

Glass Bottle
1g
96% min. (GC)
2-methyl-1h-indazole-4-boronic acid pinacol ester
258.13
258.13
2-Methyl-2H-indazole-4-boronic acid pinacol ester
145°C to 148°C
885698-95-3
C14H19BN2O2
YQMBOMPXHDRRIM-UHFFFAOYSA-N
44118254
97%
Safety and Handling

Safety and Handling

GHS Signal Word: Warning