Please login to your online account to display your discounted pricing

2-Methyl-2H-indazole-3-carbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC44404DA

 View more versions of this product

Catalog No. CC44404DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 160.18
Quantity 1g
Assay 97%
CAS 34252-54-5
Molecular Formula C9H8N2O
Assay Percent Range 97%
Chemical Name or Material 2-Methyl-2H-indazole-3-carbaldehyde
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 2-Methyl-2H-indazole-3-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8N2O
Formula Weight 160.18