missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2H-1,4-benzoxazin-3(4H)-one, 97%

Manufacturer:  Alfa Aesar™ H6107503

 View more versions of this product

Catalog No. AAH6107503



Specifications

Specifications

2-Methyl-2H-1,4-benzoxazin-3(4H)-one
21744-83-2
MFCD04141926
PILVMCZSXIINLV-UHFFFAOYSA-N
2-methyl-4H-1,4-benzoxazin-3-one
301730
97%
1g
C9H9NO2
2-methyl-2h-benzo b 1,4 oxazin-3 4h-one, 2-methyl-2h-1,4-benzoxazin-3 4h-one, 2-methyl-1,4-benzoxazin-3-one, 2-methyl-1,4-benzoxazin-3-4h-one, 2-methyl-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2-methyl-2,4-dihydro-1,4-benzoxazin-3-one, acmc-1cjvq, 2-methyl-1,4 benzoxazin-3 4h-one, 2-methyl-4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,2-methyl
CC1C(=O)NC2=CC=CC=C2O1
163.176
163.18

Chemical Identifiers

21744-83-2
163.176
PILVMCZSXIINLV-UHFFFAOYSA-N
301730
CC1C(=O)NC2=CC=CC=C2O1
C9H9NO2
MFCD04141926
2-methyl-2h-benzo b 1,4 oxazin-3 4h-one, 2-methyl-2h-1,4-benzoxazin-3 4h-one, 2-methyl-1,4-benzoxazin-3-one, 2-methyl-1,4-benzoxazin-3-4h-one, 2-methyl-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2-methyl-2,4-dihydro-1,4-benzoxazin-3-one, acmc-1cjvq, 2-methyl-1,4 benzoxazin-3 4h-one, 2-methyl-4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,2-methyl
2-methyl-4H-1,4-benzoxazin-3-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents