missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2H-1,4-benzothiazin-3(4H)-one, 97%

$55.51 - $175.80

Chemical Identifiers

CAS 7028-57-1
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD02941599
InChI Key MCEOSIYJRZMQNS-UHFFFAOYSA-N
Synonym 2-methyl-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 2-methyl, 2-methyl-2h-benzo b 1,4 thiazin-3 4h-one, 2-methyl-3,4-dihydro-2h-1,4-benzothiazin-3-one, 2-methyl-2,4-dihydro-1,4-benzothiazin-3-one, 2-methyl-4h-benzo 1,4 thiazin-3-one, 2-methyl-4h-benzo e 1,4-thiazaperhydroin-3-one
PubChem CID 3803849
IUPAC Name 2-methyl-4H-1,4-benzothiazin-3-one
SMILES CC1C(=O)NC2=CC=CC=C2S1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61012MD View Documents Alfa Aesar™
H61012MD
250mg Each for $55.51
 
AAH6101203 View Documents Alfa Aesar™
H6101203
1g Each for $175.80
 
Specifications

Chemical Identifiers

7028-57-1
179.237
MCEOSIYJRZMQNS-UHFFFAOYSA-N
3803849
CC1C(=O)NC2=CC=CC=C2S1
C9H9NOS
MFCD02941599
2-methyl-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 2-methyl, 2-methyl-2h-benzo b 1,4 thiazin-3 4h-one, 2-methyl-3,4-dihydro-2h-1,4-benzothiazin-3-one, 2-methyl-2,4-dihydro-1,4-benzothiazin-3-one, 2-methyl-4h-benzo 1,4 thiazin-3-one, 2-methyl-4h-benzo e 1,4-thiazaperhydroin-3-one
2-methyl-4H-1,4-benzothiazin-3-one

Specifications

250mg
C9H9NOS
2-methyl-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 2-methyl, 2-methyl-2h-benzo b 1,4 thiazin-3 4h-one, 2-methyl-3,4-dihydro-2h-1,4-benzothiazin-3-one, 2-methyl-2,4-dihydro-1,4-benzothiazin-3-one, 2-methyl-4h-benzo 1,4 thiazin-3-one, 2-methyl-4h-benzo e 1,4-thiazaperhydroin-3-one
CC1C(=O)NC2=CC=CC=C2S1
179.237
179.24
2-Methyl-2H-1,4-benzothiazin-3(4H)-one
7028-57-1
MFCD02941599
MCEOSIYJRZMQNS-UHFFFAOYSA-N
2-methyl-4H-1,4-benzothiazin-3-one
3803849
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning