Please login to your online account to display your discounted pricing

2-Methyl-2-vinyloxirane, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-vinyloxirane
CAS 1838-94-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C5H8O
Formula Weight 84.12
Synonym Isoprene monoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC440680250

 
thermo fisher scientific
AC440680250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC440680050

 
thermo fisher scientific
AC440680050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 84.12
Assay ≥94.0%
Physical Form Liquid
Quantity 25g
Color Colorless
Density 0.8g/mL
CAS Max % 100.0
CAS Min % 94.0
Synonym Isoprene monoxide
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
CAS 1838-94-4
Molecular Formula C5H8O
Chemical Name or Material 2-Methyl-2-vinyloxirane
Assay Percent Range 95%
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.