missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-vinyloxirane, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440680250

 View more versions of this product

Catalog No. AC440680250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methyl-2-vinyloxirane
−8°C
0.8
Colorless
1838-94-4
100.0
C5H8O
FVCDMHWSPLRYAB-UHFFFAOYSA-N
2-ethenyl-2-methyloxirane
92166
Liquid
0.8000g/mL
Authentic
80.0°C to 81.0°C
25g
94.0
94% min. (GC)
2-methyl-2-vinyl-oxirane, 2-methyl-2-vinyloxirane, 3,4-epoxy-3-methyl-1-butene, 4,4,4-3fmet-1-2-furyl-1,3-butandione, acmc-20m5m5, ccris 5685, isoprene monoxide, oxirane, 2-ethenyl-2-methyl, oxirane, 2-ethenyl-2-methyl-, 2r
CC1(CO1)C=C
84.12
84.12
95%

Chemical Identifiers

1838-94-4
84.12
2-methyl-2-vinyl-oxirane, 2-methyl-2-vinyloxirane, 3,4-epoxy-3-methyl-1-butene, 4,4,4-3fmet-1-2-furyl-1,3-butandione, acmc-20m5m5, ccris 5685, isoprene monoxide, oxirane, 2-ethenyl-2-methyl, oxirane, 2-ethenyl-2-methyl-, 2r
2-ethenyl-2-methyloxirane
C5H8O
FVCDMHWSPLRYAB-UHFFFAOYSA-N
92166
CC1(CO1)C=C
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with pl

GHS Signal Word: Danger

SDS Documents