Learn More
2-Methyl-2-thiopseudourea hemisulfate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 127891000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Methyl-2-thiopseudourea hemisulfate | |
867-44-7 | |
100.0 | |
98% | |
C2H6N2S·1/2H2SO4 | |
MFCD00013055 | |
s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1 | |
BZZXQZOBAUXLHZ-UHFFFAOYSA-N | |
methyl carbamimidothioate;sulfuric acid | |
13347 | |
98% |
98% | |
97.5 | |
Authentic | |
Plastic bottle | |
[CH3SC(=NH)NH2]2·1/2H2SO4 | |
100g | |
Solubility in water: 260g/L (20°C). Other solubilities: 4g/L methanol (20°C) | |
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O | |
139.18 | |
139.18 |
Chemical Identifiers
867-44-7 | |
139.18 | |
BZZXQZOBAUXLHZ-UHFFFAOYSA-N | |
13347 | |
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O |
C2H6N2S·1/2H2SO4 | |
MFCD00013055 | |
s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1 | |
methyl carbamimidothioate;sulfuric acid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 212-759-7
RTECSNumber : UM9200000
TSCA : TSCA
RUO â Research Use Only