Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methyl-2-pyridylamino)ethanol, 98%

$57.57 - $219.99

Chemical Identifiers

Chemical Name or Material 2-(Methyl-2-pyridylamino)ethanol
CAS 122321-04-4
Molecular Formula C8H12N2O
Formula Weight 152.19
MDL Number MFCD02667633
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2555518

  SDS
alfa aesar™
B2555518
50g Each for $57.57

AAB2555530

  SDS
alfa aesar™
B2555530
250g Each for $219.99
Description & Specifications

Specifications

Formula Weight 152.19
Chemical Name or Material 2-(Methyl-2-pyridylamino)ethanol
Assay 98%
Molecular Formula C8H12N2O
MDL Number MFCD02667633
CAS 122321-04-4
Health Hazard 1 Danger
Quantity 50g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-Methyl-2-pyridylaminoethanol is used as a reagent to synthesize Rosiglitazone (Maleate: R693500), a thiazolinedione drug used to reduce insulin resistance in patients with Type 2 diabetes.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.